Vitamin B12a hydrochloride

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydroxocobalamin hydrochloride, Hydroxocobalamin xhydrochloride

462080010 462080050
ACRO462080010EA 2550 DKK
ACRO462080010 ACRO462080050
Vitamin B12a hydrochloride
Vitamin B12a hydrochloride

  • Bioactive

Formel: C₆₂H₈₉CoN₁₃O₁₅P·HCl
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00167463
CAS-nummer: 58288-50-9

Bestil nu

Specifikationer Testresultater

Appearance Dark red to dark purple Crystalline powder or crystals
Infrared spectrum Conforms
Assay ≥85 %
Loss on drying ≤12 %
HPLC ≥90 %
Identification Chlorides: passes test
UV Lambda max 525 nm
Absorbance scale 0.31 to 0.35 (A525/A351)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR