Pirfenidone

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

464720010 464722500
ACRO464720010EA 2690 DKK
ACRO464720010 ACRO464722500
Pirfenidone
Pirfenidone

  • Bioactive

Formel: C₁₂H₁₁NO
Smeltepunkt: 109 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00866047
CAS-nummer: 53179-13-8

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 111 °C
Loss on drying ≤0.5 % (1 g, 80 °C, 4 hr, 75 kPa)
Solubility (5% in CH2Cl2) Clear colorless to yellow
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR