(+)-3-Carene
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
463990050
463990250
ACRO463990050EA
218
DKK
ACRO463990050
ACRO463990250
(+)-3-Carene
(+)-3-Carene
- Bioactive
Formel:
C₁₀H₁₆ Kogepunkt: 170 °C Massefylde: 0,865 Flammepunkt: 46 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00066417 CAS-nummer: 498-15-7 UN nummer: 2319 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4710 to 1.4760 (20°C, 589 nm) |
Density | 0.854 to 0.884 kg/l |
GC | ≥88 % |
Specific optical rotation | +13.0° to +17.0° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...