Ritalinic acid

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

463000010 463000050
ACRO463000010EA 532 DKK
ACRO463000010 ACRO463000050
Ritalinic acid
Ritalinic acid

  • Bioactive

Formel: C13H17NO2
Smeltepunkt: 245 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06200695
CAS-nummer: 19395-41-6

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR