Labelling tapes, dissolvable paper tape for thermal transfer printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
150603 150604 150605 150606 150607
BRDY150603EA 312 DKK
BRDY150603 BRDY150604 BRDY150605 BRDY150606 BRDY150607
Labelling tapes, dissolvable paper tape for thermal transfer printers
Etiketter
Continuous tapes for printers.
  • Removable acrylic adhesive
  • Matte finish
  • Autoclavable
  • Dissolves completely in water leaving no adhesive residue
  • Continuous tape is not cut to length, it gets cut by the printer
  • This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves
Water dissolvable paper labels are self-adhesive and extremely easy to apply. Exposing the label to warm water will completely dissolve it within 30 seconds, without damage to the surface it was applied to, and without leaving any residue. It reduces the labour required to remove labels, reduces maintenance to filters, and drains and enables easy re-identification of reusable items in a range of industries.
These labelling tapes should be used with IP-R4306, R4311, R4306, IP-R4302, and R4302 ribbons. They are compatible with the BBP®11 Label Printer, BBP®12 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 76mm Core Printers.