Water baths, WTB series
Leverandør: MEMMERT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WTB11 WTB15 WTB24 WTB35 WTB50 WTB6
462-0610EA 7750 DKK
462-0610 462-0611 462-0612 462-0613 462-0614 462-0615
Water baths, WTB series
Bade Vandbade, uden omrøring
Ideal for temperature control of ointments, emulsions, samples, plates and nutrient solutions in the laboratory, as well as warm storage and calibration in the industrial segment.
  • Intuitive operation, customisable remote screen CustomView
  • Practical and robust: Quick water drainage
  • Easy to clean
  • Flexible expansion with customised accessories
  • Calibration function for precise temperature settings
The WTB waterbaths are available in six model sizes (6, 11, 15, 24, 35 and 50 L). The operating concept (touchscreen) impresses with its intuitive and convenient menu navigation. A particular highlight: Thanks to the personalisable remote viewing through the CustomView, the user can always keep an eye on important information.
The high-quality stainless steel tanks with rounded corners are easy to clean and can be completely drained using the standard drain valve without leaving any residue behind. The compact ergonomic design means the WTB models fit on any laboratory bench.
Thanks to the modular concept of this modern water bath, maximum flexibility is guaranteed. If your application calls for it, a circulation pump, cooling unit, shaking device or test tube holders can easily be retrofitted at any time.
Setting temperature range: 10 to 100 °C
Temperature setting accuracy: 0,1 °C
Certificeringer: Standard units are safety-approved and bear the EAC test mark.
Leveringsinformation: WTB baths include gable cover, all other accessories must be ordered separately.
Advarsel: * RT= Ambient.