Sampling spoon curved, long handle, disposable Bio
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5379-1023 5379-0023
331-0339EA 520 DKK
331-0339 331-0340
Sampling spoon curved, long handle, disposable Bio
Skeer
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.
  • Product made from renewable raw materials
  • Bio-PE, white
  • Individually packaged and optionally sterilised, for single use
  • With curved spoon shape
  • Handle length 300 mm
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
The sampling spoon can be used in pharmaceutical applications, in the chemical industry and for food and cosmetics.