Thermal transfer printable labels, series R6400
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
120999 121003 121004 121005 121009 121010 121011 121012
BRDY120999EA 1270 DKK
BRDY120999 BRDY121003 BRDY121004 BRDY121005 BRDY121009 BRDY121010 BRDY121011 BRDY121012
Thermal transfer printable labels, series R6400
Etiketter
Polypropylene (B-7425) material is ideal for general purpose barcode applications and withstands liquid nitrogen, autoclave and other harsh environments.
  • Die-cut labels are on a roll
  • General identification, laboratory labelling and temperature indicating labelling applications
  • Service temperature: –80 to 70 °C
  • This R6400 series has matt finish
  • Thickness: 0,102 mm
  • Material with autoclavable and cryogenic storage properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.
For freezer or liquid nitrogen applications, B-7425 must be applied prior to being frozen.