Thermal transfer printable labels, series R4900
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
062368 062370 062357
BRDY062368EA 2390 DKK
BRDY062368 BRDY062370 BRDY062357
Thermal transfer printable labels, series R4900
Etiketter
Printable, nylon cloth labels for thermal transfer printers.
  • Die-cut labels are on a roll
  • Electrical and laboratory application
  • Service temperature: –40 to 90 °C
  • This R4900 series has matt finish
  • Thickness: 0,165 mm
  • Material with autoclavable and cryogenic storage properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.