Muffle furnaces with fold down door, Tmax 1100 °C and controller R7, LE series (LE 1/11 - LE24/11)
Leverandør: NABERTHERM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LE013K17N1 LE022K17N1 LE062K17N1 LE142K17N1 LE241K17N1
455-0927EA 14570 DKK
455-0927 455-0928 455-0929 455-0930 455-0934
Muffle furnaces with fold down door, Tmax 1100 °C and controller R7, LE series (LE 1/11 - LE24/11)
Ovne Muffelovne
With their convincing price/performance ratio and the fast heat-up rates, these compact muffle furnaces are perfect for many applications in the laboratory. Quality features like the dual-shell furnace housing of rust-free stainless steel and their compact, lightweight construction. The heating elements encased in quartz glass tubes make these models reliable partners for your application. The furnaces have Tmax 1100 °C, working temperature of 1050 °C.
  • Heating from two sides via heating elements protected in quartz glass tubes
  • Fast heating times
  • Exclusive use of insulation materials without categorisation according to EC Regulation No. 1272/2008 (CLP). This explicitly means that alumino silicate wool, also known as 'refractory ceramic fibre' (RCF), which is classified and possibly carcinogenic, is not used
  • Maintenance-friendly replacement of heating elements and insulation
  • Fold down door which can also be used as a work platform
These furnaces are equipped with the R7 non programmable controller. All units require a 220 to 240 V single-phase power supply.
* Temperature Uniformity of ±5 K in an empty workspace above 800 °C.
Bestillingsoplysninger: Optional accessories include chimney, chimney with fan (only for LE 6/11 - LE 24/11) or catalytic converter with fan (only for LE 6/11 - LE 24/11).
Advarsel: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table.
Refence note heat-up times: Heating time of the empty and closed furnace up to Tmax -100 K (If connected at 230 V 1/N/PE).