Immersion thermostats, MAGIO™ MS and MX
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9032201 9033201
461-1427EA 32430 DKK
461-1427 461-1428
Immersion thermostats, MAGIO™ MS and MX
Cirkulatorsystemer Åbne cirkulatorsystemer Styreenheder til termostater
The MAGIO™ series of bridge mounted circulators operate with a working temperature range of −50 to +300 °C* and a heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ immersion thermostats can be adapted to different tanks and applications.
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding external applications
  • Automatic shut off in the event of high temperature or low liquid level to avoid any potential damage
  • Bright, easy to read display
  • Powerful, continuously adjustable pressure/suction pump
  • Suitable for use with tanks up to 100 L, immersion depth 150 mm (MS-Z) or 200 mm (MX-Z)
The high resolution, modern TFT touch display has a multi-lingual user interface and shows all important information at a glance. Three large, predefined main screens clearly display data and graphics with various application priorities. All display functions are easy to operate with a fingertip.
Modern interfaces make remote control easy and practical data management possible as well as allowing integration into process structures. All MAGIO™ units have integrated PT100 connection, USB interface, RS232 and Ethernet. Analogue interfaces are available as optional accessories.
Max. flow rate / max. pressure: 31 L/min, 0,92 bar (pressure), 0,4 bar (suction).
Certificeringer: Classification III according to DIN 12876-1
IP protection class according to DIN EN 60529: IP 20
Advarsel: *For applications near or below ambient temperature a cooling coil or Julabo immersion cooler is needed.