D7000 series particulate filters, Secure Click™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D7915 D7925 D7935
111-2567EA 720 DKK
111-2567 111-2568 111-2569
D7000 series particulate filters, Secure Click™
Åndedrætsværn Genanvendelige masker Maskefiltre
3M™ Secure Click™ D7000 series particulate filters are designed for use with Secure Click™ 800 series respirators, Secure Click™ D8000 series filters and D701 filter retainer.
  • New dual-flow filters allow for easier breathing
  • Protects against particulates
  • Quick, simple connection; simply align connections and push until an audible click is heard
  • Use with 3M™ Secure Click™ 800 series reusable respirators
  • Swept-back design allows an enhanced field of view and comfort
  • Use for protection in hazardous environments
D7000 series Secure Click™ particulateMore Product Information