D8000 series cartridges, Secure Click™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D8051_U D8055_U D8094 D8095
111-2561EA 695 DKK
111-2561 111-2562 111-2564 111-2565
D8000 series cartridges, Secure Click™
Åndedrætsværn Genanvendelige masker Maskefiltre
3M™ Secure Click™ cartridges feature a new dual-flow design to allow for easier breathing.
  • New dual-flow cartridges allow for easier breathing
  • Protect against certain organic vapours with a boiling point above 65 °C
  • Quick, simple cartridge connection; simply align connections and push until an audible click is heard
  • Use with 3M™ Secure Click™ 800 series reusable respirators
  • Swept-back design allows an enhanced field of view and compatibility with welding and grinding shields
Secure Click™ A1 D8051 cartridges protect against organic gases and vapours with a boiling point above 65 °C. Suitable for use with Secure Click™ 800 series reusable respirators, it can also be used in combination with Secure-Click™ D7900 series particulate filters with a D701 filter retainer.
Secure Click™ A1B1E1K1 D8059 cartridges protect against organic gases and vapours with a boiling point above 65 °C, inorganic gases, acid gases and ammonia. Suitable for use with Secure Click™ 800 series reusable respirators, they can also be used in combination with Secure-Click™ D7900 series particulate filters with a D701 filter retainer.
Secure Click™ A2 D8055 cartridges protect against organic gases and vapours with a boiling point above 65 °C. Suitable for use with Secure Click™ 800 series reusable respirators, they can also be used in combination with Secure-Click™ D7900 series particulate filters with a D701 filter retainer.
Secure Click™ A2P3 R D8095 cartridges protects against organic vapours with a boiling point above 65 °C. They are suitable for use with Secure Click™ 800 series reusable respirators.
Secure Click™ A1B1E1K1P3 R D8094 cartridges protect against organic gases and vapours with a boiling point above 65 °C, inorganic gases, acid gases and ammonia, as well as particulates. They also offer protection against formaldehyde up to 10 ppm. They are suitable for use with Secure Click™ 800 series reusable respirators.