VWR® LTS Compatible Adjustable Volume Pipettors
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
613-6918EA 8960 DKK
613-6918 613-6919 613-6920 613-6921 613-6922 613-6923 613-6924 613-6925 613-6926 613-6927 613-6928 613-6929 613-6930 613-6931 613-6932
VWR® LTS Compatible Adjustable Volume Pipettors
Pipetter Automatiske pipetter
The VWR® LTS compatible pipettors reduce ejection force with a unique positive stop technology. This technology along with lighter plunger springs, a durable light weight design, and a comfortable handle design with perfect symmetry makes this the best choice for a lab trying to improve ergonomics.
  • Fully compatible with Rainin LTS tips
  • Lighter springs give a more comfortable and controlled draw and dispense
  • Constructed with durable lightweight materials
  • The ejector is cushioned to reduce dynamic stress on your thumb
  • Precision piston design produces accurate and precise measurements with low channel to channel variability in multichannel
  • Hitting the positive stop ensures that you've made a secure seal on both single and multichannel, no more guessing or reattaching tips
The VWR® LTS compatible pipettor will help improve the ergonomics in your lab by reducing the highest force in the pipetting cycle, ejection force. The unique positive stop technology works with LTS compatible pipette tips to produce an accurate measurement using lighter forces.
Certificeringer: ISO 8655 certified. Calibration certificate included in the box. Unique serial ID number under the finger hook for GLP/GMP compliance.
Advarsel: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.