VWR®, Laboratory Bottles, Coated
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-4267EA 271 DKK
215-4267 215-4269 215-4271
VWR®, Laboratory Bottles, Coated
Flasker Glasflasker
Lavet af ravgult borsilikat 3,3 glas med gennemsigtig plastbelægning med lang levetid. Belægningen beskytter flasken mod ridser, splinter og lækkende væsker.
  • Very good chemical resistance
  • Autoclaving or steam sterilisation at 121 or 134 °C/20 minutes
  • Frequent autoclaving reduces splintering protection
  • Long-term temperature resistance up to 140 °C/>30 minutes
  • Amber coloured, ideal for use with light-sensitive liquids
  • Delivered with mounted pouring ring and blue PP screw cap
The economical alternative for a wide range of laboratory applications. Purely external application of the colour ensures unchanged glass properties on the inner surface of the bottles.