VWR® Reagent Bottles, Narrow Neck
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-4365EA 1360 DKK
215-4365 215-4366 215-4367 215-4368 215-4369 215-4370 215-4372 215-4373 215-4374 215-4375 215-4376 215-4377 215-4371
VWR® Reagent Bottles, Narrow Neck
Flasker Glasflasker
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
  • Good chemical and physical properties
  • Clear or amber glass, delivered with black PP screw cap
  • Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Certificeringer: Thread in accordance with DIN 168.