Stopcocks for carboys
Leverandør: hünersdorff GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
520100 520700 520705 520105
216-5291EA 159 DKK
216-5291 216-5292 HUEN520705 HUEN520105
Stopcocks for carboys
Stophaner
Made of fracture-proof and food-safe polyethylene with sleeve connection for a hose.
  • Very resistant to acids, alkalis and other chemicals
  • allow for extra-precise dosing