Disposable glass conical centrifuge tubes, ungraduated, without screw cap, PYREX®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
99502-10 99502-50
734-4251EA 967 DKK
734-4251 734-4254
Disposable glass conical centrifuge tubes, ungraduated, without screw cap, PYREX®
Rør Centrifugerør
Corning disposable glass centrifuge tubes are manufactured from PYREX® Type I borosilicate glass.
These tubes feature more consistent uniform wall thickness, a well-formed conical tapered tip and durable tip radius. The optimally shaped tip design allows for easy removal of samples following centrifugation. The tubes are designed to withstand centrifugation up to 3000 RCF and are packed in convenient, easy to use packs, which are shrink sealed for protection during shipment.
Bestillingsoplysninger: All caps must be ordered separately. For compatible screw caps, select either disposableMore Product Information