General purpose gloves, DermiFlex Plus Glove, A351
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A351G8RM A351G8RL A351G8RXL
PRTWA351G8RMEA 463 DKK
PRTWA351G8RM PRTWA351G8RL PRTWA351G8RXL
General purpose gloves, DermiFlex Plus Glove, A351
Handsker
Dotted palm for enhanced grip and added palm protection.
  • Lightweight and comfortable
  • Retail tag which aids presentation for retail sales
  • 15 gauge liner for extra dexterity
Breathable seamless lining ideal for precision handling in dry environments.
Elastane dipping: Nitrile foam
Certificeringer: Standard: AS/NZS 2161 .2, AS/NZS 2161 .3, ANSI/ISEA 105: 2016 CUT Level A1, EN388:2016 +A1:2018 - 4.1.3.1.X., EN 420.