Safety shoes, lace-up, Steelite™ ULTRA Safety Loire Low Cut Trainer, FW36
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW36GRR42 FW36GRR43 FW36GRR44 FW36GRR45 FW36GRR46 FW36GRR47 FW36GRR39 FW36GRR40 FW36GRR41
PRTWFW36GRR42EA 525 DKK
PRTWFW36GRR42 PRTWFW36GRR43 PRTWFW36GRR44 PRTWFW36GRR45 PRTWFW36GRR46 PRTWFW36GRR47 PRTWFW36GRR39 PRTWFW36GRR40 PRTWFW36GRR41
Safety shoes, lace-up, Steelite™ ULTRA Safety Loire Low Cut Trainer, FW36
Sko og støvler
Anti-static safety footwear made from cow suede leather plus a dual density light weight EVA/rubber sole, resistant to heat (300 °C).
  • Protective steel toecap
  • Steel midsole
  • Energy absorbing seat region
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
Certificeringer: AS 2210.3; EN ISO 20345:2011, SRC, S1P, HRO.