Safety ankle boots, lace-up, Eden
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FD15BKR42 FD15BKR38 FD15BKR39 FD15BKR40 FD15BKR41 FD15BKR43 FD15BKR44 FD15BKR45 FD15BKR46 FD15BKR47 FD15BKR48
PRTWFD15BKR42EA 930 DKK
PRTWFD15BKR42 PRTWFD15BKR38 PRTWFD15BKR39 PRTWFD15BKR40 PRTWFD15BKR41 PRTWFD15BKR43 PRTWFD15BKR44 PRTWFD15BKR45 PRTWFD15BKR46 PRTWFD15BKR47 PRTWFD15BKR48
Safety ankle boots, lace-up, Eden
Sko og støvler
Anti-static ankle boots made from full-grain leather upper plus an PU/rubber dual-density sole, resistant to oil and heat (300 °C). Provides complete protection, the full grain leather upper provides a barrier to all weather conditions. High lace up style with a robust zip for a comfortable fit and reliable support in rugged environments.
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
Injected PU/Rubber outsole for supreme grip and energy absorption.
Colour: Black
Certificeringer: EN ISO 20345, AS 2210.3, S3, HRO, CI, HI, FO