Safety spectacles, Pan View, PW38
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PW38BKR PW38CLR PW38COR PW38SOR
PRTWPW38BKREA 27.1 DKK
PRTWPW38BKR PRTWPW38CLR PRTWPW38COR PRTWPW38SOR
Safety spectacles, Pan View, PW38
Briller
These sleek and sporty style glasses are designed for safety and comfort. The Pan View features lightweight construction and soft touch temples. The single wrap-around lens offers protection from the sides and front.
  • 99% UV protection
  • One piece curved lens with moulded nose-bridge
  • Soft arm tips and rubber nose-bridge for ultra comfort
  • Anti-scratch coating for added durability
  • Anti-fog coating for additional comfort to the wearer
  • Retail bag which aids presentation for retail sales
  • Free spec cord
Certificeringer: EN 166 1 FT; EN 170 2C-1.2 (CLEAR); EN 172 5-3.1 (SMOKE); ANSI/ISEA Z87 + (Clear); ANSI/ISEA Z87 + U6 L3 (Smoke)
Leveringsinformation: Individually packed for vending machines.