Bizflame™ FR21 Overalls, Flame Resistant, Anti-Static
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FR21NARXXL FR21NARM FR21NARL FR21NARXL FR21NARXXXL FR21NAR4XL FR21NARXS FR21NARS FR21NAR5XL
PRTWFR21NARXXLEA 890 DKK
PRTWFR21NARXXL PRTWFR21NARM PRTWFR21NARL PRTWFR21NARXL PRTWFR21NARXXXL PRTWFR21NAR4XL PRTWFR21NARXS PRTWFR21NARS PRTWFR21NAR5XL
Bizflame™ FR21 Overalls, Flame Resistant, Anti-Static
Arbejdstøj
These overalls are perfect for the extra warm weather demands of the offshore industry. Constructed with a lighter weight highly innovative flame-resistant twill fabric.
  • Flame resistant industrial wash tape
  • Two tier knee pad pockets allowing two positioning options
  • Two-way zip for quick and easy access
  • Hook and loop cuffs for a secure fit
  • Radio loops for easy clipping of a radio
  • Ten pockets for ample storage
These provides protection against radiant, convective and contact heat and guaranteed flame resistance for life of garment.
Shell fabric: Bizflame Plus (210 g)
Certificeringer: EN 1149 -5; EN ISO 11612 (A1+A2, B1, C1, F1); EN ISO 11611 Class 1 (A1+A2).
Class 2 Welding Protection.