Cut resistant gloves, A621
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A621BKRS A621BKRXXL A621BKRL A621BKRXL A621BKRM
PRTWA621BKRSEA 635 DKK
PRTWA621BKRS PRTWA621BKRXXL PRTWA621BKRL PRTWA621BKRXL PRTWA621BKRM
Cut resistant gloves, A621
Handsker
Cut resistance combined with superb grip in oily conditions. ³/₄ nitrile foam dipping absorbs oil keeping hands clean and dry to ensure secure handling.
  • ANSI cut level A3
  • Materials: HDPE, glass fibre, nitrile foam
  • 100% breathable seamless liner
  • 13 gauge liner for a perfect fit
  • Level D cut resistance with abrasion and tear resistance
Nitrile foam coating for excellent grip in wet and dry conditions.
Certificeringer: Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .4, ANSI/ISEA 105: 2016 CUT Level A4, EN388:2016 +A1:2018 - 4.X.4.3.D., EN 407 X.1.X.X.X.X., EN 420.