Safety waistcoats, KS63 Xenon Toolvest
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KS63YERM KS63YERL KS63YERXL KS63YERXXL
PRTWKS63YERMEA 391 DKK
PRTWKS63YERM PRTWKS63YERL PRTWKS63YERXL PRTWKS63YERXXL
Safety waistcoats, KS63 Xenon Toolvest
Arbejdstøj
The multi-pocket toolvest combines modern design with highly durable, fluorescent fabric.
  • Shell Fabric: Kingsmill 245 g
  • Reflective tape for increased visibility
  • 10 pockets for ample storage
  • Adjustable waist for a perfect fit
  • Radio loop for easy clipping of a radio
The toolvest is crammed with features including twin stitched seams, Dual ID holder, holster pockets and waist belt for comfortable fit and support when bending and stretching.
These garments are made from a premium, heavyweight 300 g polyester/cotton fabric combined with a high tensile polyester for reinforcement in the high wear areas. The bright fluorescent yellow fabric combined with the retro reflective stripes offer the highest possible visibility in poor light or adverse weather conditions.
Certificeringer: EN ISO 20471 Class 2; ANSI standards.
Advarsel: Wash with like colours. Do not dry clean.