Gloves, vinyl, A905
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A905BLUM A905BLUL A905BLUXL
PRTWA905BLUMEA 63.6 DKK
PRTWA905BLUM PRTWA905BLUL PRTWA905BLUXL
Gloves, vinyl, A905
Handsker
These disposable gloves are manufactured from high grade poly-vinyl chloride.
  • Latex-free
  • Powder-free
  • Smooth and non textured with a beaded cuff
  • DOP free
  • Super soft for superior user comfort
  • Stretchy to reduce hand fatigue
Ideal for food handling and processing, general work and domestic use.
Certificeringer: Cat. I, Food contact approved