VWR®, Total Inorganic Carbon (TIC) Standards
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88248.180EA
754
DKK
88248.180
88248.260
88249.180
88249.260
VWR®, Total Inorganic Carbon (TIC) Standards
Standarder
Miljø analyse standarder
Standard solutions of sodium carbonate.
1000 mg/L sodium carbonate solution according to EN 1484.
Certificeringer: Manufacturer quality system is ISO 9001 certified.
Accredited according to ISO/IEC 17025 - Testing and to ISO 17034.