Reusable earplugs, flexible fit, E-A-R™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
328-1001 328-1000
111-2511EA 0 DKK
111-2511 111-2510
Reusable earplugs, flexible fit, E-A-R™
Ørepropper Genanvendelige ørepropper
E-A-R™ Flexible Fit HA earplugs have a patented earplug foam formulation which makes them fully washable and reusable. The soft foam tip and flexible fitting stem allows easy insertion into the ear using one or two hands, as the foam tip conforms to the unique shape of the ear canal to help block out noise. These earplugs are ideal for activities that require gloves or cause dirty hands, or situations when only one hand is free to fit the earplugs.
  • Firm, yet flexible stem for one- or two- hand insertion
  • Soft foam stem for secure grip
  • No roll-down of soft foam tip helps keep ear tip clean
  • Good for workers with gloved or dirty hands
  • Foam encapsulated construction
  • Single Number Rating (SNR) 35 dB (two-hand insertion) and 30 dB (one-hand insertion)
  • Compatible with the 3M™ E-A-Rfit™ Dual-Ear Validation System
The shape of the earplug tip, along with the soft conforming foam, helps to provide a snug yet gentle fit that reduces exposure to hazardous noise levels. The no roll-down design of the foam tip keeps ear tips clean when hands are dirty. Single Number Rating (SNR) is 35 dB when tested using two-hand insertion method, and 30 dB, when tested using one-hand insertion method. Fit test probes are available to conduct fit testing using the 3M™ E-A-Rfit™ Dual Ear Validation System.
The patented design and innovative thermoplastic materials make this earplug one of the most versatile and durable 'push to fit' style earplugs. Workplace applications include assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding. machine operations, painting, sanding and welding. These earplugs are appropriate for use in automotive, construction, general manufacturing, metal fabrication, military maintenance, repair and operation (MRO), mining, oil and gas, pharmaceutical and transportation industries.