Safety helmet, SecureFit™ X5500 Series
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
X5501V-CE-4 7100175598
MMMAX5501V-CE-4EA 778 DKK
MMMAX5501V-CE-4 MMMA7100175598
Safety helmet, SecureFit™ X5500 Series
Hjelme Sikkerhedshjelme
The SecureFit™ X5501V-CE safety helmet includes an easy-to-adjust, 4-point ratchet suspension system that features the exclusive 3M™ Pressure Diffusion Technology. This brimless, white safety helmet has a UVicator™ sensor that indicates when the hard hat has been overexposed to UV-radiation and requires replacement.
  • Secure, comfortable head protection against small falling objects and general industrial use
  • Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
  • 4-point ratchet suspension allows customised height and fit with multiple levels of vertical adjustment
  • Vents allow airflow through the helmet
  • Inspired by climbing helmets with a brimless design to provide a clear upward view
Inspired by modern climbing helmets, the SecureFit™ safety helmet X5501V-CE offers protection against small falling objects. Vents allow airflow through the helmet and the height positioning on the head is adjustable. The easy-to-adjust, 4-point ratchet suspension system allows smooth tightening of the headband as needed. This suspension includes the exclusive 3M™ Pressure Diffusion Technology, which not only provides a secure fit but, by reducing up to 20% of forehead pressure compared to conventional helmet suspensions, also helps improve comfort.
This white helmet features a proprietary 3M™ UVicator™ sensor that changes from red to white when exposed to damaging levels of ultraviolet (UV) light. As UV rays can cause hard hat shells to become brittle and compromise their protection level, the sensor changing colour indicates that it’s time to replace the helmet. Scotchlite™ Reflective Material can also be added to increase visibility under low light conditions.
Accessory slots and clips are designed to be compatible with a variety of face shields, earmuffs and other accessories, and an optional chin strap with a magnetic or plastic buckle can be added.
Certificeringer: Approved for lateral deformation and meets the EN 397 standard for general industrial use.