Safety prescription spectacles, Solus™ 2000 series
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S2020AF-BLU S2015AF-BLU S2025AF-BLU
111-2577EA 194 DKK
111-2577 111-2576 111-2578
Safety prescription spectacles, Solus™ 2000 series
Briller
Comfortable, stylish eyewear with a vast array of colour and lens tint options, the Solus™ 2000 prescription series provides eye protection with a stylish flare.
  • Adjustable ratchet temples
  • Readers with anti-fog/anti-scratch coating
  • Available in a range of diopters: +1,5, +2,0, or +2,5
  • Polycarbonate lenses absorb 99,9% of UVA and UVB rays
The Solus™ 2000 prescription series features adjustable ratchet temples that allow the wearer to customise the fit. Choose from several lens coating options, including Scotchgard™ anti-fog, multiple lens colours, and features like a removable foam gasket and diopters to create the best protective eyewear solution.
Certificeringer: Compliant with EN 166, EN 170; W EN 166 F CE (frame); 2C-1.2 3M 1 FT CE (lens)