Earmuffs, LiteCom headsets, Peltor™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MT53H7P3E4400E MT53H7B4400-EU MT53H7A4400-EU
MMMAMT53H7P3E4400EEA 8050 DKK
MMMAMT53H7P3E4400E MMMAMT53H7B4400-EU MMMAMT53H7A4400-EU
Earmuffs, LiteCom headsets, Peltor™
Høreværn Høreværn med radio
Peltor™ LiteCom Headsets are hearing protectors with a built-in, pre-programmed two-way communication radio and an integrated noise cancelling microphone. They are built for demanding environments to help users communicate efficiently over hazardous noise.
  • Hearing protector with built-in analogue two-way communication radio
  • Eight channels in the PMR 446 MHz band
  • 38 sub-channels
  • Noise cancelling waterproof microphone provides voice-operated transmission (VOX) for hands-free use
  • Voice guided menu system
  • Auto power off function
  • Clear battery status indication
  • Communication radio in the PMR 446 MHz frequency range
Peltor™ LiteCom Headsets facilitate clear communicate while keeping user's hearing protected in noisy environments. These headsets have a built-in, pre-programmed two-way communication radio, which allows headset-to-headset conversation. Use eight channels in the PMR 446 MHz band, while 38 sub-channels let users communicate individually using the same channel. The noise cancelling microphone provides voice-operated transmission for hands-free use.