Bottles, UN approved, with self-sealing screw cap
Leverandør: LAMAPLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
452 453 451
LAMA452EA 40 DKK
LAMA452 LAMA453 LAMA451
Bottles, UN approved, with self-sealing screw cap
Flasker Plastflasker
Sturdy HDPE bottles are supplied with tamper-evident screw caps.
  • Robust HDPE bottles
  • Good chemical resistance
  • UN approved for liquids
  • Leakproof design
Bottles are ideal for sample shipping.