Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated High Temperature Fused Silica
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HI99-05-DHT-1EA 184 DKK
HI99-05-DHT-1 HI99-05-DHT-2 HI99-05-DHT-5 HI99-05-DHT-10 HI99-10-DHT-1 HI99-10-DHT-2 HI99-10-DHT-5 HI99-10-DHT-10 HI99-10-DHT-20 HI99-15-DHT-1 HI99-15-DHT-2 HI99-15-DHT-5 HI99-15-DHT-10 HI99-15-DHT-20 HI99-15-DHT-30 HI99-18-DHT-1 HI99-18-DHT-2 HI99-18-DHT-5 HI99-18-DHT-10 HI99-18-DHT-20 HI99-18-DHT-30 HI99-20-DHT-1 HI99-20-DHT-2 HI99-20-DHT-5 HI99-20-DHT-10 HI99-20-DHT-20 HI99-20-DHT-30 HI99-25-DHT-1 HI99-25-DHT-2 HI99-25-DHT-5 HI99-25-DHT-10 HI99-25-DHT-20 HI99-25-DHT-30 HI99-25-DHT-60 HI99-32-DHT-1 HI99-32-DHT-2 HI99-32-DHT-5 HI99-32-DHT-10 HI99-32-DHT-20 HI99-32-DHT-30 HI99-32-DHT-60 HI99-53-DHT-1 HI99-53-DHT-2 HI99-53-DHT-5 HI99-53-DHT-10 HI99-53-DHT-20 HI99-53-DHT-30 HI99-53-DHT-60
Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated High Temperature Fused Silica
Kromatografikolonner
HMDS deactivated high temperature fused silica retention gap / precolumn. Deactivation mainly for apolar solvents.