Avantor® Hichrom, Retention Gap/Precolumn, DPTMDS Deactivated Fused Silica
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HI95-05-D-1EA 168 DKK
HI95-05-D-1 HI95-05-D-2 HI95-05-D-5 HI95-05-D-10 HI95-10-D-1 HI95-10-D-2 HI95-10-D-5 HI95-10-D-10 HI95-10-D-20 HI95-15-D-1 HI95-15-D-2 HI95-15-D-5 HI95-15-D-10 HI95-15-D-20 HI95-15-D-30 HI95-18-D-1 HI95-18-D-2 HI95-18-D-5 HI95-18-D-10 HI95-18-D-20 HI95-18-D-30 HI95-20-D-1 HI95-20-D-2 HI95-20-D-5 HI95-20-D-10 HI95-20-D-20 HI95-20-D-30 HI95-25-D-1 HI95-25-D-2 HI95-25-D-5 HI95-25-D-10 HI95-25-D-20 HI95-25-D-30 HI95-25-D-60 HI95-32-D-1 HI95-32-D-2 HI95-32-D-5 HI95-32-D-10 HI95-32-D-20 HI95-32-D-30 HI95-32-D-60 HI95-53-D-1 HI95-53-D-2 HI95-53-D-5 HI95-53-D-10 HI95-53-D-20 HI95-53-D-30 HI95-53-D-60
Avantor® Hichrom, Retention Gap/Precolumn, DPTMDS Deactivated Fused Silica
Kromatografikolonner
DPTMDS deactivated fused silica retention gap / precolumn. General purpose deactivation for both apolar and polar solvents.