Test kits, COD cuvette tests for salt water
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LCK1814 LCK1914 LCK1714
705-1167EA 1030 DKK
705-1167 705-1168 HACHLCK1714
Test kits, COD cuvette tests for salt water
Miljø test kit
Applicable for COD testing in water samples with high chloride concentrations up to 20000 mg/L.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
  • No cumbersome sample pre-treatment required
Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packed in a fully equipped box.