Cobalt AAS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E4017-100ML E4017-500ML E4018-100ML E4018-500ML
HONCE4017-100MLEA 262 DKK
HONCE4017-100ML HONCE4017-500ML HONCE4018-100ML HONCE4018-500ML
Cobalt AAS Standards, (CRMs), Fluka™
Standarder Kationiske standarder Cobalt Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificeringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).