Nitrile gloves, Showa 6112PF
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6112PFEBTL 6112PFEBTM 6112PFEBTS 6112PFEBTXL 6112PFEBTXS 6112PFEBTXXL
BEST6112PFEBTLEA 221 DKK
BEST6112PFEBTL BEST6112PFEBTM BEST6112PFEBTS BEST6112PFEBTXL BEST6112PFEBTXS BEST6112PFEBTXXL
Nitrile gloves, Showa 6112PF
Handsker
These versatile, single-use gloves provide sensitivity and dexterity. The 0,10 mm thick nitrile is also waterproof and provides extreme comfort during long periods of use.
  • Lightweight, unsupported
  • Powder-free
  • Latex-free to prevent allergic reactions
  • Smooth grip
By using SHOWA's Eco Best Technology ®, the 6112PF's are completely eco-friendly, breaking down by 82% in just 386 days when tested in a laboratory.
Approved for food handling and contact, these gloves are ideal for food preparation, use in laboratories, and general repair or maintenance work.
Certificeringer: EN ISO 374-1:2016 Type B (KPT); EN ISO 374-5:2016 VIRUS; Cat. III
Food contact approved. GreenCircle certified. ASTM D5526
Emballering: 100 gloves per dispenser. The dispenser is made of 85% recycled cardboard, the outer carton is made of 90% recycled material.