Labels, dissolvable paper, for J2000 printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
151239 151241 151243 151244 151240
BRDY151239EA 1180 DKK
BRDY151239 BRDY151241 BRDY151243 BRDY151244 BRDY151240
Labels, dissolvable paper, for J2000 printers
Etiketter
Rectangular dissolvable paper labels are an easy to remove, environmentally responsible, print-on-demand inkjet label solution. Ideal for use in laboratory settings, asset tracking, or general use.
  • Removable acrylic adhesive
  • Matte finish
  • Maximum service temperature: 172 °C
  • Minimum service temperature: –40 °C
  • Uniform 0,094 mm thickness
The water dissolvable paper label is self-adhesive and extremely easy to apply. Exposing the label to warm water will completely dissolve it within 30s, without damage to the surface it was applied to, and without leaving any residue. It reduces the labour required to remove labels, reduces maintenance to filters and drains, and enables easy re-identification of reusable items in a wide range of industries.