Avantor® Hichrom HI-POF 1, GC Columns
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HI55-18-018-20EA 8210 DKK
HI55-18-018-20 HI55-25-015-30 HI55-25-015-60 HI55-25-025-15 HI55-25-025-30 HI55-25-025-60 HI55-32-025-30 HI55-32-032-30 HI55-32-050-15 HI55-32-050-30 HI55-53-050-30
Avantor® Hichrom HI-POF 1, GC Columns
Kromatografikolonner
Application specifc columns developed for pesticides, herbicides, insecticides analysis etc.
Max temperature limits may change depending on the film thickness.