Absorbent pads and rolls, Universal Plus
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
813842 813843 813846 834177 813847
BRDY813842EA 1170 DKK
BRDY813842 BRDY813843 BRDY813846 BRDY834177 BRDY813847
Absorbent pads and rolls, Universal Plus
Spild materialer
These absorbent pads and rolls are used when chemicals and hazardous liquids are involved.
  • 2-ply meltblown and spunbound on one side, dimpled and perforated
  • Absorb a wide range of chemicals, including acids and caustics
  • One-sided spunbond gives extra strength as well as lower lint
  • Perforated for flexibility – tear at the perforations for smaller sized pads
Chemically inert product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid. Green colour alerts workers that product is being used for hazardous liquids. Green colourMore Product Information