Labels for BBP3x/S3xxx printers, preprinted
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
142037 142040 142043 142044 142045 142046 142047
BRDY142037EA 3540 DKK
BRDY142037 BRDY142040 BRDY142043 BRDY142044 BRDY142045 BRDY142046 BRDY142047
Labels for BBP3x/S3xxx printers, preprinted
Etiketter
For quickly creating NFPA and colour bar labels.
  • Pre-printed right-to-know chemical labels
  • Allows the printer to produce five colour chemical labels in a single print pass
  • Indoor/outdoor grade material withstands exposure to moisture, chemicals and handling
  • Color: Blue/red/yellow on white
Made up of vinyl.