Labels for thermal transfer printers, type B-484
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
062391 062390
BRDY062391EA 1890 DKK
BRDY062391 BRDY062390
Labels for thermal transfer printers, type B-484
Etiketter
Printable, permanent polyester labels for thermal transfer printers. Ultra-aggressive adhesive designed for powder coated surfaces, on angled and curved surfaces.
  • Fuels or oils resistant
  • High and low temperatures resistant
  • Solvents or chemicals resistant
  • Friction resistant
  • Extra strong adhesive
Certificeringer: UL recognised. Compliant with Directive 2011/65/EU and 2015/863.