Ultra-Thin Electrostatic Dissipative (ESD) Polyimide Labels for B33 Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
133870 133874 133878 133798 153173 153165 153160 153166
BRDY133870EA 3830 DKK
BRDY133870 BRDY133874 BRDY133878 BRDY133798 BRDY153173 BRDY153165 BRDY153160 BRDY153166
Ultra-Thin Electrostatic Dissipative (ESD) Polyimide Labels for B33 Printers
Etiketter
Static dissipative polyimide (B-718) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • Service temperature of –70 to +100 °C
Low profile, high temperature, glossy white polyimide material features a static dissipative adhesive and static dissipative release liner. Surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008.
Certificeringer: UL Approved; meets requirements of MIL-STD-202G, method 215K.