3-Hydroxy-2-nitropyridin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol
A14887.09 A14887.18 A14887.22 A14887.30
A14887.09EA 421 DKK
A14887.09 A14887.18 A14887.22 A14887.30
3-Hydroxy-2-nitropyridin ≥98%
3-Hydroxy-2-nitropyridine
Purity: 98% CAS No.: 15128-82-2 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: 69-73° Beilstein Reference: 124473
Formel: C₅H₄N₂O₃
Molekylvægt: 140.1 g/mol
Smeltepunkt: 69…73 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006258
CAS-nummer: 15128-82-2
EINECS: 239-191-2