Chemical resistant overalls, AlphaTec® 2000 Standard, Model 111
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GR20T-00111-03 GR20T-00111-07 GR20T-00111-02 GR20T-00111-04 GR20T-00111-06 GR20T-00111-05 WH20T-00111-08 WH20T-00111-09
ANSEGR20T-00111-03EA 4890 DKK
ANSEGR20T-00111-03 ANSEGR20T-00111-07 ANSEGR20T-00111-02 ANSEGR20T-00111-04 ANSEGR20T-00111-06 ANSEGR20T-00111-05 ANSEWH20T-00111-08 ANSEWH20T-00111-09
Chemical resistant overalls, AlphaTec® 2000 Standard, Model 111
Engangsbeklædning
Superior breathable microporous laminate technology, offering Type 5/6 protection.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone-free, critical in spray painting applications
Coveralls feature a 3-piece hood, 2-way front zipper with resealable storm flap, and finger loops. Elasticated hood, waist, wrists, and ankles. Garments are manufactured from non-woven microporous polyethylene laminate, with bound seams.
Recommended for cleanroom cleaning and preparation, production line support and maintenance, maintenance of plant and machines, and veterinary services.
Certificeringer: Type 5-B; Type 6-B; EN 1073-2; EN 14126; DIN 32781; EN 1149-5; Category III
Emballering: Individually packaged; 40 pieces/carton.