Ferrocen ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Packaged under argon.
87202.18
87202.30
87202.A1
87202.18EA
474
DKK
87202.18
87202.30
87202.A1
Ferrocen ≥99%
Ferrocen
Purity: 99%
CAS No.: 102-54-5
Molecular Formula: C10H10Fe
Formula Weight: 186.04
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Yellow to orange
Odour: Characteristic
Melting Point: 173-174°
Boiling Point: 249°
Merck Reference: 14,4037
UN No.: 1325
MDL No.: MFCD00001427
Formel:
C₁₀H₁₀Fe Kogepunkt: 249 °C Smeltepunkt: 173…174 °C Massefylde: 1,49 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001427 CAS-nummer: 102-54-5 EINECS: 203-039-3 UN nummer: 1325 ADR: 4.1,II Merck Index: 14,4037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...