Tetraethylammoniumnitrat 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

420310250 420310100
ACRO420310250EA 1180 DKK
ACRO420310250 ACRO420310100
Tetraethylammoniumnitrat 99%
Tetraethylammonium nitrate
Formel: C₈H₂₀N₂O₃
Smeltepunkt: >240 °C
Massefylde: 1,1622 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041978
CAS-nummer: 1941-26-0
EINECS: 217-725-5
UN nummer: 1479
ADR: 5.1,II

Bestil nu

Specifikationer Testresultater

Appearance White to yellowish-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR