Dodecyltrimethylammoniumbromid ≥99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide
A10761.09 A10761.18 A10761.30
A10761.09EA 613 DKK
A10761.09 A10761.18 A10761.30
Dodecyltrimethylammoniumbromid ≥99%
Dodecyltrimethylammonium bromide
Purity: 99% CAS No.: 1119-94-4 Molecular Formula: CH3(CH2)11N(CH3)3Br Formula Weight: 308.35 Melting Point: ca 245° dec. Beilstein Reference: 3597463 UN No.: 3077 MDL No.: MFCD00011767
Formel: C₁₅H₃₄BrN
Molekylvægt: 308.35 g/mol
Smeltepunkt: ∼ 245 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011767
CAS-nummer: 1119-94-4
EINECS: 214-290-3
ADR: 9 III