tert-Butylchlordimethylsilan 98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
183930050 183930250 183931000 183935000
ACRO183930050EA 314 DKK
ACRO183930050 ACRO183930250 ACRO183931000 ACRO183935000
tert-Butylchlordimethylsilan 98%
tert-Butylchlordimethylsilan
Formel: C₆H₁₅ClSi
Kogepunkt: 125 °C (1013 hPa)
Smeltepunkt: 85…91 °C
Massefylde: 0,87 g/cm³
Flammepunkt: 22 °C
MDL Number: MFCD00000501
CAS-nummer: 18162-48-6
EINECS: 242-042-4
UN nummer: 2921
Bestil nu
Specifikationer Testresultater
Appearance Colorless to white Crystals
Infrared spectrum Conforms
Melting point 85 to 91 °C
GC ≥97.5 %
HPLC ≥97.5 %