Industrial gloves, HyFlex® 11-738
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-738/11 11-738/8 11-738/10 11-738/9 11-738/6 11-738/7
ANSE11-738/11EA 2530 DKK
ANSE11-738/11 ANSE11-738/8 ANSE11-738/10 ANSE11-738/9 ANSE11-738/6 ANSE11-738/7
Industrial gloves, HyFlex® 11-738
Handsker
These tough gloves feature a unique combination of INTERCEPT Technology® yarn, Nylon, Lycra®, and glass fiber.
  • Ultra strong fibers provide extreme resistance against cuts and burrs
  • Reinforced thumb crotch for increased protection & extended use life
  • INTERCEPT Cut Resistance Technology for extreme cut protection
  • Made with water-based polyurethane for enhanced comfort and dexterity
  • Silicone- and solvent-free
FORTIX™ Technology for extended product use life and increased protection with up to 12x more durability in the thumb crotch region versus unreinforced glove equivalents.
Recommended for: Handling of sharp-edged objects, pre-assembling, cutting of small dry or lightly oiled parts and maintenance; handling of glass sheets, sharpedged objects; handling of metal sheets, cutting of dry, painted or galvanised pieces.
Certificeringer: EN 388:2016 (4X42C); Cat. II
Oeko-Tex® 100 Certified
Emballering: 12 pairs per bag.